Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myristic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tetradecanoic acid Tetradecylic acid

A12067-0E A12067-36 A12067-0E A12067-30 A12067-30 A12067-36
AAA12067-0EEA 138 USD InStock
AAA12067-0E AAA12067-36 AAAA12067-30 CAAAAA12067-30 CAAAA12067-36 CAAAA12067-0E
Myristic acid 98%
Myristic acid


Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 326 °C
Melting Pt: 53…56 °C
Density: 0,862
Flash Pt: 175 °C(347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002744
CAS Number: 544-63-8
EINECS: 208-875-2
Merck Index: 12,06416
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty