Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Perfluoroheptanoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Tridecafluoroenanthic acid Tridecafluoroheptanoic acid Tridecafluoroheptylic acid Perfluoroenanthic acid

A12092-14 A12092-06 A12092-06 A12092-14
AAA12092-14EA 194.99 USD InStock
AAA12092-14 AAA12092-06 CAAAA12092-14 CAAAA12092-06
Perfluoroheptanoic acid 98+%
Perfluoroheptanoic acid


Formula: C₇HF₁₃O₂
Boiling Pt: 89 °C/30 mm
Melting Pt: 31…36 °C (lit.)
Density: 1.792
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039604
CAS Number: 375-85-9
EINECS: 206-798-9
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty