Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromobutyryl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A12094-22 A12094-22 A12094-14 A12094-06 A12094-06 A12094-14
AAA12094-06EA 39.09 USD InStock
CAAAA12094-22 AAA12094-06 AAA12094-22 CAAAA12094-14 AAA12094-14 CAAAA12094-06
4-Bromobutyryl chloride 97%
4-Bromobutyryl chloride


Formula: C₄H₆BrClO
Boiling Pt: 102…103 °C/40 mm
Density: 1.600
Flash Pt: 91 °C(195 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000753
CAS Number: 927-58-2
EINECS: 213-154-0
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty