Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: N,N,N',N'-Tetramethyl-1,4-phenylenediamine dihydrochloride TMPD 2 HCl

A12107-09 A12107-09 A12107-18 A12107-18 A12107-30 A12107-30
AAA12107-30EA 1033.99 USD InStock
AAA12107-30 AAA12107-09 CAAAA12107-18 AAA12107-18 CAAAA12107-09 CAAAA12107-30
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride


Formula: C₁₀H₁₈Cl₂N₂
MW: 237.17 g/mol
Melting Pt: ca 228 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012482
CAS Number: 637-01-4
EINECS: 211-274-8
Merck Index: 12,09367
https://us-legacy.vwr.com/stibo/web/std.lang.all/61/62/2346162.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty