Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Chloroacetophenone 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: p-Chloroacetophenone

A12115-30 A12115-0B A12115-0I A12115-0B A12115-30 A12115-0I
AAA12115-0IEA 962.75 USD InStock
AAA12115-0I CAAAA12115-0I AAA12115-30 CAAAA12115-0B AAA12115-0B CAAAA12115-30
4'-Chloroacetophenone 98+%
4'-Chloroacetophenone


Formula: C₈H₇ClO
MW: 154.6 g/mol
Boiling Pt: 230…232 °C
Melting Pt: 17…20 °C
Density: 1.190
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000624
CAS Number: 99-91-2
EINECS: 202-800-7
ADR: 6.1,II
Merck Index: 13,02133
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty