Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Dodecanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Dodecyl alcohol 1-Hydroxy dodecane Undecyl carbinol

A12228-0I A12228-30 A12228-0B A12228-0B A12228-0I A12228-30
AAA12228-30EA 35.01 USD InStock
CAAAA12228-0B AAA12228-30 AAA12228-0I CAAAA12228-0I AAA12228-0B CAAAA12228-30
1-Dodecanol 98%
1-Dodecanol


Formula: H₃C(CH₂)₁₀CH₂OH
MW: 186.34 g/mol
Boiling Pt: 258…260 °C
Melting Pt: 24…27 °C
Density: 0.830
Flash Pt: 127 °C(260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004753
CAS Number: 112-53-8
EINECS: 203-982-0
UN: 3077
ADR: "9",III
Merck Index: 13,03439
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty