Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Piperidone 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: δ-Valerolactame

A12271-36 A12271-22 A12271-14 A12271-14 A12271-36 A12271-22
AAAA12271-36EA 1067.79 USD InStock
AAAA12271-36 CAAAA12271-14 CAAAAA12271-36 CAAAA12271-22 AAA12271-22 AAA12271-14
2-Piperidone 98%
2-Piperidone


Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 255…257 °C
Melting Pt: 35…39 °C
Density: 1,073 g/cm³ (40 °C)
Flash Pt: 147 °C(296 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006037
CAS Number: 675-20-7
EINECS: 211-622-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty