Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dibenzothiophene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dibenzo[b,d]thiophene

A12288-14 A12288-36 A12288-14 A12288-22 A12288-22 A12288-36
AAA12288-22EA 160.99 USD InStock
CAAAA12288-22 AAA12288-22 AAA12288-14 AAA12288-36 CAAAA12288-14 CAAAA12288-36
Dibenzothiophene 98%
Dibenzothiophene


Formula: C₁₂H₈S
MW: 184.26 g/mol
Boiling Pt: 332…333 °C
Melting Pt: 97…100 °C
Density: 1.1295 g/cm³ (125 °C)
Flash Pt: 170 °C(338 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004969
CAS Number: 132-65-0
EINECS: 205-072-9
UN: 3077
ADR: "9",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty