Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethoxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals

A12309-22 A12309-36 A12309-14 A12309-22 A12309-36 A12309-14
AAA12309-36EA 318.99 USD InStock
CAAAA12309-22 AAA12309-36 AAA12309-14 CAAAA12309-36 CAAAA12309-14 AAA12309-22
4-Ethoxybenzoic acid 98+%
4-Ethoxybenzoic acid


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Melting Pt: 197…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00002545
CAS Number: 619-86-3
EINECS: 210-616-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty