Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calcium benzoate hydrate, (max. 8% H₂O) 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Benzoic acid calcium salt

A12358-36 A12358-36 A12358-0E A12358-0E A12358-22 A12358-22
AAA12358-36EA 173.99 USD InStock
CAAAA12358-22 CAAAA12358-36 AAA12358-36 CAAAA12358-0E AAA12358-0E AAA12358-22
Calcium benzoate hydrate, (max. 8% H₂O) 98%
Calcium benzoate hydrate
Calcium benzoate hydrate is used in used in the food industry as a preservative.


Formula: C₁₄H₁₀CaO₄
MW: 282.31 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00167164
CAS Number: 2090-05-3
EINECS: 218-235-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty