Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl carbazate 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: tert-Butyl hydrazinecarboxylate

A12383-36 A12383-22 A12383-14 A12383-22 A12383-36 A12383-14
AAAA12383-36EA 593.99 USD InStock
CAAAA12383-22 AAAA12383-36 CAAAA12383-14 AAA12383-14 CAAAAA12383-36 AAA12383-22
tert-Butyl carbazate 98+%
tert-Butyl carbazate


Formula: C₅H₁₂N₂O₂
MW: 132.16 g/mol
Boiling Pt: 63…65 °C/0.1 mm
Melting Pt: 37…41 °C
Flash Pt: 91 °C(195 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00007593
CAS Number: 870-46-2
EINECS: 212-795-3
UN: 2926
ADR: "4.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty