Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphoenolpyruvic acid monocyclohexylammonium salt 98%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-(Phosphonooxy)-2-propenoic acid monocyclohexylammonium saltPEP·CHXPhospho(enol)pyruvic acid cyclohexylammonium salt

A12385-14 A12385-03 A12385-06 A12385-14 A12385-03
AAAA12385-06EA 311.99 USD InStock
CAAAA12385-03 CAAAA12385-14 AAAA12385-06 AAA12385-14 AAA12385-03
Phosphoenolpyruvic acid monocyclohexylammonium salt 98%
Phosphoenolpyruvic acid monocyclohexylammonium salt


Formula: C₃H₅O₆P
MW: 267.22 g/mol
Melting Pt: ca 140 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00036375
CAS Number: 10526-80-4
EINECS: 234-084-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty