Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodoacetic acid 97+%

Supplier: Thermo Scientific Chemicals
Danger

A12410-14 A12410-22 A12410-14 A12410-22
AAA12410-22EA 258.67 USD InStock
AAA12410-22 AAA12410-14 CAAAA12410-14 CAAAA12410-22
Iodoacetic acid 97+%
Iodoacetic acid
As a reagent for the modification of sulfhydryl groups


Formula: C₂H₃IO₂
MW: 185.95 g/mol
Boiling Pt: 208 °C (dec.)
Melting Pt: 79…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002685
CAS Number: 64-69-7
EINECS: 200-590-1
UN: 2923
ADR: "8",III
Merck Index: 13,05047
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty