Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Difluorophenyl isocyanate 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2,4-Difluorophenylisocyanate

A12415-14 A12415-06 A12415-06 A12415-22 A12415-14 A12415-22
AAA12415-06EA 48.49 USD InStock
AAA12415-06 AAA12415-14 AAA12415-22 CAAAA12415-14 CAAAA12415-06 CAAAA12415-22
2,4-Difluorophenyl isocyanate 98+%
2,4-Difluorophenyl isocyanate


Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Boiling Pt: 64…66 °C/10 mm
Density: 1.310
Flash Pt: 50 °C(122 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001997
CAS Number: 59025-55-7
UN: 2478
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty