Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Tribromophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A12424-30 A12424-0B
AAA12424-0BEA 140.99 USD InStock
AAA12424-0B AAA12424-30
2,4,6-Tribromophenol 98%
2,4,6-Tribromophenol


Formula: C₆H₃Br₃O
MW: 330.8 g/mol
Boiling Pt: 282…290 °C (subl.)
Melting Pt: 86…92 °C
Density: 2.550
Storage Temperature: Ambient
MDL Number: MFCD00002150
CAS Number: 118-79-6
EINECS: 204-278-6
UN: 3077
ADR: "9",III
Merck Index: 12,09744
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty