Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Benzyloxy)benzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Benzyloxybenzoic acid

A12425-30 A12425-09 A12425-09 A12425-18 A12425-30 A12425-18
AAA12425-18EA 305.99 USD InStock
CAAAA12425-09 CAAAA12425-18 AAA12425-18 CAAAA12425-30 AAA12425-30 AAA12425-09
2-(Benzyloxy)benzoic acid 98%
2-(Benzyloxy)benzoic acid


Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00051940
CAS Number: 14389-86-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty