Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclobutanol 96%

Supplier: Thermo Scientific Chemicals
Danger

A12430-06 A12430-03 A12430-06 A12430-14 A12430-03 A12430-14
AAAA12430-14EA 1181.99 USD InStock
AAAA12430-14 AAA12430-03 CAAAA12430-06 CAAAAA12430-14 CAAAA12430-03 AAA12430-06
Cyclobutanol 96%
Cyclobutanol


Formula: C₄H₈O
Boiling Pt: 123…124 °C
Density: 0.925
Flash Pt: 21 °C(69 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001318
CAS Number: 2919-23-5
EINECS: 220-858-1
UN: 1987
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty