Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Homovanillic acid 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: (4-Hydroxy-3-methoxyphenyl)acetic acid

A12441-14 A12441-06 A12441-03 A12441-06 A12441-03 A12441-14
AAAA12441-14EA 735.99 USD InStock
AAAA12441-14 AAA12441-03 CAAAA12441-06 AAA12441-06 CAAAAA12441-14 CAAAA12441-03
Homovanillic acid 98+%
Homovanillic acid
Fluorimetric reagent and major catecholamine metabolite


Formula: (C₆H₃(OH)(OCH₃))CH₂COOH
MW: 182.18 g/mol
Melting Pt: 140…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00004350
CAS Number: 306-08-1
EINECS: 206-176-7
Merck Index: 13,04761
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty