Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2-nitrobenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A12493-14 A12493-06 A12493-22 A12493-14 A12493-06 A12493-22
AAA12493-06EA 19.79 USD InStock
CAAAA12493-14 CAAAA12493-22 AAA12493-06 AAA12493-14 CAAAA12493-06 AAA12493-22
5-Chloro-2-nitrobenzoic acid 99%
5-Chloro-2-nitrobenzoic acid


Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Melting Pt: 136…140 °C
Density: 1,593 g/cm³ (18 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007290
CAS Number: 2516-95-2
EINECS: 219-738-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty