Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Glutamic acid hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S)-2-Aminopentanedioic acid hydrochloride

A12505-0B A12505-30 A12505-0B A12505-30
AAA12505-0BEA 80.59 USD InStock
AAA12505-0B AAA12505-30 CAAAA12505-0B CAAAA12505-30
L(+)-Glutamic acid hydrochloride 99%
L(+)-Glutamic acid hydrochloride


Formula: C₅H₁₀ClNO₄
MW: 183.59 g/mol
Melting Pt: 203 °C (dec.)
Density: 1.525
Storage Temperature: Ambient
MDL Number: MFCD00012619
CAS Number: 138-15-8
EINECS: 205-315-9
Order Now
Specification Test Results
Optical rotation +23° (c=6 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty