Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω-Bromo-4-chloroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 4-Chlorophenacyl bromide 2-Bromo-4'-chloroacetophenone α-Bromo-4-chloroacetophenone Bromomethyl p-chlorophenyl ketone

A12607-36 A12607-14 A12607-36 A12607-22 A12607-14 A12607-22
AAA12607-14EA 46.29 USD InStock
CAAAA12607-22 CAAAA12607-36 CAAAA12607-14 AAA12607-14 AAA12607-36 AAA12607-22
ω-Bromo-4-chloroacetophenone 98%
ω-Bromo-4-chloroacetophenone


Formula: C₈H₆BrClO
MW: 233.49 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00000625
CAS Number: 536-38-9
EINECS: 208-631-5
UN: 3261
ADR: "8",III
Merck Index: 13,02172
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty