Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyrylthiocholine iodide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2-Butyrylthioethyl)trimethylammonium iodide (Propylcarbonylthioethyl)trimethylammonium iodide (2-(Butyrylthio)ethyl)trimethylammonium iodide

A12622-14 A12622-14 A12622-06 A12622-06
AAA12622-06EA 48.49 USD InStock
CAAAA12622-14 AAA12622-06 AAA12622-14 CAAAA12622-06
Butyrylthiocholine iodide 98%
Butyrylthiocholine iodide


Formula: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
MW: 317.23 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011845
CAS Number: 1866-16-6
EINECS: 217-475-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty