Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromo-4-methoxybenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12653-06 A12653-14 A12653-14 A12653-06
AAA12653-06EA 80.09 USD InStock
AAA12653-06 CAAAA12653-06 CAAAA12653-14 AAA12653-14
3-Bromo-4-methoxybenzoic acid 98+%
3-Bromo-4-methoxybenzoic acid


Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 217…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00020295
CAS Number: 99-58-1
EINECS: 202-768-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty