Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Butanesulfonic acid sodium salt 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium 1-butansulphonate

A12654-22 A12654-14 A12654-22 A12654-06 A12654-06 A12654-14
AAA12654-06EA 50.99 USD InStock
AAA12654-06 CAAAA12654-06 CAAAA12654-14 CAAAA12654-22 AAA12654-22 AAA12654-14
1-Butanesulfonic acid sodium salt 99%
1-Butansulphonic acid sodium salt


Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1
EINECS: 219-201-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty