Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1S)-(-)-Borneol 97+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (1S-endo)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol L(-)-Borneol

A12684-30 A12684-18 A12684-18 A12684-0B A12684-0B A12684-30
AAA12684-30EA 183.18 USD InStock
CAAAA12684-0B AAA12684-30 AAA12684-0B CAAAA12684-18 CAAAA12684-30 AAA12684-18
(1S)-(-)-Borneol 97+%
(1S)-(-)-Borneol


Formula: C₁₀H₁₈O
MW: 154.25 g/mol
Boiling Pt: 210 °C
Melting Pt: 208…211 °C
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003759
CAS Number: 464-45-9
EINECS: 207-353-1
UN: 1312
ADR: "4.1",III
Merck Index: 13,01328
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty