Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-3-hydroxypyridine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-3-pyridinol

A12687-14 A12687-22 A12687-22 A12687-06 A12687-06 A12687-14
AAA12687-14EA 216.99 USD InStock
CAAAA12687-06 AAA12687-14 AAA12687-22 AAA12687-06 CAAAA12687-14 CAAAA12687-22
2-Bromo-3-hydroxypyridine 99%
2-Bromo-3-hydroxypyridine


Formula: C₅H₄BrNO
MW: 174 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0
EINECS: 229-547-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty