Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Amantadine hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Adamantylamine hydrochloride 1-Aminoadamantane hydrochloride 1-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride 1-Adamantanammonium chloride Tricyclo[3.3.1.1(3,7)]dec-1-ylamine hydrochloride

A12699-36 A12699-14 A12699-14 A12699-22 A12699-36 A12699-22
AAA12699-22EA 209 USD InStock
AAA12699-22 AAA12699-14 AAA12699-36 CAAAA12699-22 CAAAA12699-36 CAAAA12699-14
Amantadine hydrochloride 99%
Amantadine hydrochloride
Antiviral drug that inhibits ion channels of influenza and disrupts T-cell development


Formula: C₁₀H₁₈ClN
MW: 187.71 g/mol
Melting Pt: 360 °C
Storage Temperature: Ambient
MDL Number: MFCD00074723
CAS Number: 665-66-7
EINECS: 211-560-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty