Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-6-nitrobenzonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Chloro-2-nitrobenzonitrile

A12703-14 A12703-06 A12703-06 A12703-14
AAA12703-06EA 76.49 USD InStock
AAA12703-06 AAA12703-14 CAAAA12703-06 CAAAA12703-14
2-Chloro-6-nitrobenzonitrile 98%
2-Chloro-6-nitrobenzonitrile


Formula: C₇H₃ClN₂O₂
MW: 182.57 g/mol
Melting Pt: 118…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007203
CAS Number: 6575-07-1
EINECS: 229-496-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty