Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anthranilic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12715-0I
AAA12715-0IEA 327.99 USD InStock
AAA12715-0I
Anthranilic acid 98+%
Anthranilic acid


Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 145…148 °C
Density: 1.412
Flash Pt: 150 °C(302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007712
CAS Number: 118-92-3
EINECS: 204-287-5
Merck Index: 13,00421
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty