Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9,10-Anthraquinone 98+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Anthraquinone

A12735-30 A12735-0B A12735-0B A12735-30
AAA12735-0BEA 215.99 USD InStock
AAA12735-0B CAAAA12735-30 AAA12735-30 CAAAA12735-0B
9,10-Anthraquinone 98+%
9,10-Anthraquinone


Formula: C₁₄H₈O₂
MW: 208.22 g/mol
Boiling Pt: 377 °C
Melting Pt: 283…287 °C
Density: 1.438
Flash Pt: 185 °C(365 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001188
CAS Number: 84-65-1
EINECS: 201-549-0
Merck Index: 13,00692
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty