Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxy-β-nitrostyrene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Methoxy-4-(2-nitrovinyl)benzenetrans-4-Methoxy-β-nitrostyrene

A12745-06 A12745-22 A12745-14 A12745-06 A12745-22 A12745-14
AAA12745-14EA 346.99 USD InStock
CAAAA12745-06 AAA12745-14 CAAAA12745-14 AAA12745-06 AAAA12745-22 CAAAAA12745-22
4-Methoxy-β-nitrostyrene 98%
4-Methoxy-β-nitrostyrene


Formula: C₉H₉NO₃
MW: 179.173 g/mol
Melting Pt: 86…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00024826
CAS Number: 3179-10-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty