Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Isopropoxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

A12760-03 A12760-06 A12760-14 A12760-03 A12760-14 A12760-06
AAA12760-03EA 38.09 USD InStock
AAA12760-03 AAA12760-14 AAA12760-06 CAAAA12760-03 CAAAA12760-14 CAAAA12760-06
4-Isopropoxybenzaldehyde 97%
4-Isopropoxybenzaldehyde


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 108…110 °C/5 mm
Storage Temperature: Ambient
MDL Number: MFCD00052357
CAS Number: 18962-05-5
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty