Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Acetyl-5-cyanothiophene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Acetylthiophene-2-carbonitrile

A12765-02 A12765-02
AAA12765-02EA 48.49 USD InStock
CAAAA12765-02 AAA12765-02
2-Acetyl-5-cyanothiophene 97%
2-Acetyl-5-cyanothiophene


Formula: C₇H₅NOS
Boiling Pt: 95…97 °C/0.5 mm
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00052314
CAS Number: 88653-55-8
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty