Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Difluoroacetic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

A12787-14 A12787-14 A12787-06 A12787-06
AAA12787-14EA 328.69 USD InStock
CAAAA12787-06 CAAAA12787-14 AAA12787-14 AAA12787-06
Difluoroacetic acid 98%
Difluoroacetic acid


Formula: C₂H₂F₂O₂
MW: 96.03 g/mol
Boiling Pt: 132…134 °C
Melting Pt: –1 °C
Density: 1.526
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004220
CAS Number: 381-73-7
EINECS: 206-839-0
UN: 3265
ADR: "8",III
Order Now
Specification Test Results
Refractive index 1.3430

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty