Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Phenyliodide

A12788-18 A12788-0B A12788-22 A12788-22 A12788-0B A12788-18
AAA12788-0BEA 403.99 USD InStock
CAAAA12788-18 AAA12788-0B CAAAAA12788-22 AAAA12788-22 CAAAA12788-0B AAA12788-18
Iodobenzene 98%
Iodobenzene


Formula: C₆H₅I
MW: 204.01 g/mol
Boiling Pt: 188…189 °C
Melting Pt: –31 °C
Density: 1,826
Flash Pt: 74 °C(165 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001029
CAS Number: 591-50-4
EINECS: 209-719-6
Merck Index: 13,05051
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty