Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Dodecansulfonic acid sodium salt 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 1-dodecansulphonate

A12829-03 A12829-14 A12829-09 A12829-09 A12829-14 A12829-03
AAA12829-14EA 388.99 USD InStock
AAA12829-14 AAA12829-03 AAAA12829-09 CAAAAA12829-09 CAAAA12829-03 CAAAA12829-14
1-Dodecansulfonic acid sodium salt 99%
1-Dodecansulphonic acid sodium salt


Formula: C₁₂H₂₅NaO₃S
MW: 272.38 g/mol
Melting Pt: >300 °C
Density: 1.218 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007527
CAS Number: 2386-53-0
EINECS: 219-200-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty