Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxy-p-anisaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-Hydroxy-4-methoxybenzaldehydeIsovanillinIsovanicalineIsovanillic aldehyde

A12866-14 A12866-06 A12866-22 A12866-14 A12866-06 A12866-22
AAA12866-14EA 68.49 USD InStock
AAA12866-14 CAAAA12866-14 CAAAA12866-22 AAAA12866-06 AAA12866-22 CAAAAA12866-06
3-Hydroxy-p-anisaldehyde 98%
3-Hydroxy-p-anisaldehyde


Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 179 °C/15 mm
Melting Pt: 112…116 °C
Density: 1,20
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0
EINECS: 210-694-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty