Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Iodotrimethylsilane 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trimethyliodosilane Trimethylsilyl iodide

Stabilization: Stabilised with copper

A12902-14 A12902-14 A12902-22 A12902-06 A12902-06 A12902-30 A12902-30 A12902-22
AAAA12902-22EA 317.42 USD InStock
AAAA12902-22 CAAAAA12902-22 CAAAAA12902-06 AAAA12902-14 CAAAAA12902-14 AAA12902-30 AAAA12902-06 CAAAA12902-30
Iodotrimethylsilane 97% stabilized
Iodotrimethylsilane


Formula: C₃H₉ISi
MW: 200.09 g/mol
Boiling Pt: 105…106 °C
Melting Pt: < 0 °C
Density: 1.440
Flash Pt: -2 °C(28 °F)
Storage Temperature: Freezer
MDL Number: MFCD00001028
CAS Number: 16029-98-4
EINECS: 240-171-0
UN: 2924
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty