Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

9-Fluorenone 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 9H-Fluoren-9-one

A12910-0E A12910-36 A12910-36 A12910-0E A12910-22 A12910-22
AAA12910-22EA 58.19 USD InStock
AAA12910-22 CAAAA12910-22 CAAAA12910-0E CAAAA12910-36 AAA12910-0E AAA12910-36
9-Fluorenone 98+%
9-Fluorenone


Formula: C₁₃H₈O
MW: 180.21 g/mol
Boiling Pt: 342 °C
Melting Pt: 82…85 °C
Density: 0.9
Flash Pt: 163 °C(325 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001141
CAS Number: 486-25-9
EINECS: 207-630-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty