Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(3-Chloro-4-fluorophenyl)indole 98%

Supplier: Thermo Scientific Chemicals
Warning

A12920-03 A12920-06 A12920-03 A12920-06
AAA12920-03EA 102.99 USD InStock
CAAAA12920-03 AAA12920-03 CAAAA12920-06 AAA12920-06
2-(3-Chloro-4-fluorophenyl)indole 98%
2-(3-Chloro-4-fluorophenyl)indole


Formula: C₁₄H₉ClFN
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00068179
CAS Number: 1868-88-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty