Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Nitroindole 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 6-Nitro-1H-indole

A12927-14 A12927-06 A12927-14 A12927-06 A12927-03 A12927-03
AAA12927-06EA 380.99 USD InStock
AAA12927-06 AAA12927-14 AAA12927-03 CAAAA12927-06 CAAAA12927-14 CAAAA12927-03
6-Nitroindole 97%
6-Nitroindole


Formula: C₈H₆N₂O₂
MW: 162.15 g/mol
Melting Pt: 141…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00051497
CAS Number: 4769-96-4
EINECS: 225-311-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty