Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluorobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: p-Fluorobenzoyl chloride

A12946-14 A12946-14 A12946-22 A12946-36 A12946-36 A12946-22
AAA12946-14EA 50 USD InStock
CAAAA12946-14 CAAAA12946-36 AAA12946-14 AAA12946-36 AAA12946-22 CAAAA12946-22
4-Fluorobenzoyl chloride 98%
4-Fluorobenzoyl chloride


Formula: C₇H₄ClFO
MW: 158.56 g/mol
Boiling Pt: 193 °C
Melting Pt: 10…12 °C
Density: 1.326
Flash Pt: 82 °C(179 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000684
CAS Number: 403-43-0
EINECS: 206-961-4
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty