Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Basic fuchsin

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Basic fuchsin hydrochloride(4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride Diamondfuchsin Rosaniline hydrochloride

A12952-14 A12952-06 A12952-22 A12952-14 A12952-06 A12952-22
AAA12952-06EA 38.44 USD InStock
AAA12952-06 AAA12952-14 CAAAA12952-06 AAA12952-22 CAAAA12952-14 CAAAA12952-22
Basic fuchsin
Basic Fuchsin
Biological stain for distinguishing coli and aerogene bacteria


Formula: C₂₀H₂₀ClN₃
MW: 337.85 g/mol
Melting Pt: 268…270 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012569
CAS Number: 632-99-5
EINECS: 211-189-6
Merck Index: 12,05686
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty