Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl isobutyrate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2-Methylpropionic acid ethyl ester

A12966-AP A12966-AE A12966-0F A12966-AP A12966-AE A12966-0F
AAA12966-0FEA 292.99 USD InStock
AAA12966-0F CAAAA12966-AP AAA12966-AE CAAAA12966-AE CAAAA12966-0F AAA12966-AP
Ethyl isobutyrate 98%
Ethyl isobutyrate


Formula: C₆H₁₂O₂
MW: 116.16 g/mol
Boiling Pt: 108…110 °C
Melting Pt: –88 °C
Density: 0.869
Flash Pt: 13 °C(55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009165
CAS Number: 97-62-1
EINECS: 202-595-4
UN: 2385
ADR: "3",II
Merck Index: 13,03847
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty