Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Iodohexadecane 98% stabilized

Supplier: Thermo Scientific Chemicals

Stabilization: Stabilised with copper

A12977-14 A12977-22 A12977-30
AAA12977-14EA 43.09 USD InStock
AAA12977-14 AAA12977-22 AAA12977-30
1-Iodohexadecane 98% stabilized
1-Iodohexadecane


Formula: C₁₆H₃₃I
MW: 352.34 g/mol
Boiling Pt: 206…207 °C/10 mm
Melting Pt: 21…23 °C
Density: 1.121
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001089
CAS Number: 544-77-4
EINECS: 208-879-4
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty