Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-4-Bromomandelic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (±)-4-Bromophenylglycolic acid

A12978-14 A12978-06 A12978-14 A12978-06 A12978-22 A12978-22
AAA12978-14EA 199.99 USD InStock
CAAAA12978-22 AAA12978-14 AAA12978-06 AAA12978-22 CAAAA12978-14 CAAAA12978-06
(±)-4-Bromomandelic acid 98+%
(±)-4-Bromomandelic acid


Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Boiling Pt: 344 °C (1013 hPa)
Melting Pt: 115…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00004232
CAS Number: 6940-50-7
EINECS: 230-085-1
Merck Index: 13,01409
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty