Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

β-D-Maltose octaacetate 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)-5-(((2R,3R,4S,5R,6R)-3,4,5-triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2,3,4-triyl triacetate

A12988-14 A12988-06 A12988-14 A12988-06 A12988-03 A12988-03
AAA12988-14EA 396.99 USD InStock
AAA12988-14 AAA12988-03 AAA12988-06 CAAAA12988-14 CAAAA12988-03 CAAAA12988-06
β-D-Maltose octaacetate 98%
beta-D-Maltose octaacetate


Formula: C₂₈H₃₈O₁₉
MW: 678.60 g/mol
Melting Pt: 157…160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079432
CAS Number: 22352-19-8
Order Now
Specification Test Results
Optical rotation +65° (c=10 in chloroform)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty