Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3-Diaminonaphthalene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: DAN2,3-Naphthalenediamine2,3-naphthylenediamine

A12993-06 A12993-03 A12993-14 A12993-06 A12993-14 A12993-03
AAA12993-06EA 443.99 USD InStock
AAA12993-06 AAA12993-14 AAA12993-03 CAAAA12993-06 CAAAA12993-03 CAAAA12993-14
2,3-Diaminonaphthalene 97%
2,3-Diaminonaphthalene


Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 193…196 °C
Density: 1.096
Storage Temperature: Ambient
MDL Number: MFCD00004116
CAS Number: 771-97-1
EINECS: 212-241-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty