Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-ethylbenzene 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(Bromophenyl)ethane

A13013-30 A13013-18 A13013-09 A13013-30 A13013-18 A13013-09
AAA13013-18EA 208.99 USD InStock
CAAAA13013-18 AAA13013-18 CAAAAA13013-30 AAAA13013-30 AAA13013-09 CAAAA13013-09
1-Bromo-4-ethylbenzene 99%
1-Bromo-4-ethylbenzene


Formula: C₈H₉Br
MW: 185.06 g/mol
Boiling Pt: 203…204 °C
Melting Pt: –43,5 °C
Density: 1,341
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000111
CAS Number: 1585-07-5
EINECS: 216-439-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty