Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-β-D-glucosamine 98+%

Supplier: Thermo Scientific Chemicals
Synonyms: 2-Acetamido-2-deoxy-D-glucoseD-GlcNAc

A13047-09 A13047-22 A13047-18 A13047-18 A13047-22 A13047-09
AAAA13047-22EA 135.99 USD InStock
CAAAAA13047-22 AAAA13047-22 CAAAA13047-18 AAA13047-18 AAA13047-09 CAAAA13047-09
N-Acetyl-β-D-glucosamine 98+%
N-Acetyl-β-D-glucosamine


Formula: C₈H₁₅NO₆
MW: 221.21 g/mol
Melting Pt: 213 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00136044
CAS Number: 7512-17-6
EINECS: 231-368-2
Order Now
Specification Test Results
Optical rotation +42° (c=2 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty